Learn More
CPC Scientific H-DPhe-DPhe-DNle-DArg-NH-4Picolyl (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109980
Encompass
SEQUENCE: H-DPhe-DPhe-DNle-DArg-NH-4Picolyl (trifluoroacetate salt)ONE-LETTER SEQUENCE: ff-DNle-r-NH4PicolylMOLECULAR FORMULA: C36H49N9O4MOLECULAR WEIGHT: 671.83STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: PainREFERENCES:SKU(s): OPIO-008A
Catalog No. 50-210-9980 Supplier CPC Scientific Supplier No. OPIO008A
SEQUENCE: H-DPhe-DPhe-DNle-DArg-NH-4Picolyl (trifluoroacetate salt)ONE-LETTER SEQUENCE: ff-DNle-r-NH4PicolylMOLECULAR FORMULA: C36H49N9O4MOLECULAR WEIGHT: 671.83STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: PainREFERENCES:SKU(s): OPIO-008A