Learn More
CPC Scientific H-DPhe-His-Leu-Leu-Arg-Glu-Val-Leu-Glu-Nle-Ala-Arg-Ala-Glu-Gln-Leu-Ala-Gln-Glu-Ala-His-Lys-Asn-Arg-Lys-Leu-Nle-Glu-Ile-Ile-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CRFS002B
Encompass
CRF receptor antagonist.ONE-LETTER SEQUENCE: DPhe-HLLREVLE-Nle-ARAEQLAQEAHKNRKL-Nle-EII-NH2MOLECULAR FORMULA: C161H271N49O43MOLECULAR WEIGHT:3581.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: Hormonal
Catalog No. 50-210-9162