Learn More
Medchemexpress LLC DPI 201-106 | 97730-95-5 | 99.5% | 466.57 | 1 ML
SDP
Catalog No. 5000400811
Encompass_Preferred
DPI 201-106 is a cardiotonic agent known for its synergistic sarcolemmal and intracellular mechanism of action. It cardioselectively modulates voltage-gated sodium channels, leading to a positive inotropic effect.
  • Cardiotonic agent
  • Synergistic sarcolemmal and intracellular mechanism of action
  • Cardioselective modulation of voltage-gated sodium channels
  • Results in a positive inotropic effect
Catalog No. 50-004-00811 Supplier Medchemexpress LLC Supplier No. HY1966610MM/1ML
DPI 201-106 is a cardiotonic agent known for its synergistic sarcolemmal and intracellular mechanism of action. It cardioselectively modulates voltage-gated sodium channels, leading to a positive inotropic effect.
  • Cardiotonic agent
  • Synergistic sarcolemmal and intracellular mechanism of action
  • Cardioselective modulation of voltage-gated sodium channels
  • Results in a positive inotropic effect