Learn More
Medchemexpress LLC DPI 201-106 (SDZ 201106) | 97730-95-5 | 99.5% | 466.57 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY19666
Encompass_Preferred
DPI 201-106 (SDZ 201106) is a cardiotonic agent that works with a synergistic sarcolemmal and intracellular mechanism of action. It demonstrates cardioselective modulation of voltage-gated sodium channels (VGSCs), leading to a positive inotropic effect. This product is for research use only and is not sold to patients.
  • Cardiotonic agent
  • Synergistic sarcolemmal and intracellular mechanism of action
  • Cardioselective modulation of voltage-gated sodium channels (VGSCs)
  • Positive inotropic effect
Catalog No. 50-004-02631