Learn More
Medchemexpress LLC Dpm-1001 | 1471172-27-6 | 99.8% | 10 MG
SDP
Catalog No. 5000233893
Encompass_Preferred
DPM-1001 is a potent, orally active, and non-competitive inhibitor of protein-tyrosine phosphatase 1B (PTP1B) that shows in vitro activity (IC50 ≈ 100 nM) and in vivo efficacy in diet-induced obese mice.
  • Potent PTP1B inhibition with IC50 around 100 nM
  • Orally active and non-competitive mechanism of action
  • Demonstrated efficacy in animal models improving glucose tolerance and reducing body weight
  • High reported purity suitable for research use
  • Available in small pack sizes for preclinical studies
Catalog No. 50-002-33893 Supplier Medchemexpress LLC Supplier No. HY12151510MG
DPM-1001 is a potent, orally active, and non-competitive inhibitor of protein-tyrosine phosphatase 1B (PTP1B) that shows in vitro activity (IC50 ≈ 100 nM) and in vivo efficacy in diet-induced obese mice.
  • Potent PTP1B inhibition with IC50 around 100 nM
  • Orally active and non-competitive mechanism of action
  • Demonstrated efficacy in animal models improving glucose tolerance and reducing body weight
  • High reported purity suitable for research use
  • Available in small pack sizes for preclinical studies