Learn More
Medchemexpress LLC Dpm-1001 | 1471172-27-6 | 99.8% | 5 MG
SDP
Catalog No. 5000232698
Encompass_Preferred
DPM-1001 is a small-molecule, orally bioavailable, non-competitive inhibitor of protein-tyrosine phosphatase 1B (PTP1B) with a reported IC50 of approximately 100 nM. It is used in preclinical research to investigate insulin and leptin signaling and metabolic regulation.
  • Non-competitive inhibitor of PTP1B (IC50 ≈ 100 nM).
  • Orally bioavailable in preclinical models.
  • Supports research into insulin and leptin signaling pathways.
  • High reported purity suitable for biochemical assays.
  • Stored cold to maintain stability and activity.
Catalog No. 50-002-32698 Supplier Medchemexpress LLC Supplier No. HY1215155MG
DPM-1001 is a small-molecule, orally bioavailable, non-competitive inhibitor of protein-tyrosine phosphatase 1B (PTP1B) with a reported IC50 of approximately 100 nM. It is used in preclinical research to investigate insulin and leptin signaling and metabolic regulation.
  • Non-competitive inhibitor of PTP1B (IC50 ≈ 100 nM).
  • Orally bioavailable in preclinical models.
  • Supports research into insulin and leptin signaling pathways.
  • High reported purity suitable for biochemical assays.
  • Stored cold to maintain stability and activity.