Learn More
AdipoGen DPM
SDP
Catalog No. 502561278
Encompass_Preferred
Chemical. CAS 143503-03-1. Formula C20H16N2O5. MW 364.35. Synthetic. Nonfluorescent N-substituted maleimides react selectively with thiol compounds to form fluorescent derivatives. Fluorescent intensity of the dimethoxyphthalimidyl group is higher than that of the nonsubsituted phthalimidyl group. The analytical sensitivity of N-4-5,6-Dimethoxy-N-phthalimidylphenylmaleimide DPM is about 3-times higher than that of N-4-2-phthalimidylphenylmaleimide PPM. DPM can be used as a precolumn labeling reagent in HPLC. DPM was also used for the determination of physiologically important thiols in biological fluids or therapeutic drug monitoring.
Catalog No. 50-256-1278 Supplier AdipoGen Supplier No. CDXD0049M250
Chemical. CAS 143503-03-1. Formula C20H16N2O5. MW 364.35. Synthetic. Nonfluorescent N-substituted maleimides react selectively with thiol compounds to form fluorescent derivatives. Fluorescent intensity of the dimethoxyphthalimidyl group is higher than that of the nonsubsituted phthalimidyl group. The analytical sensitivity of N-4-5,6-Dimethoxy-N-phthalimidylphenylmaleimide DPM is about 3-times higher than that of N-4-2-phthalimidylphenylmaleimide PPM. DPM can be used as a precolumn labeling reagent in HPLC. DPM was also used for the determination of physiologically important thiols in biological fluids or therapeutic drug monitoring.