Learn More
Apexbio Technology LLC DPN 1428-67-7 5mg
SDP
Catalog No. 5000089286
Encompass_Preferred
DPN (1428-67-7) is a non-steroidal agonist targeting estrogen receptor (ER ) It is designed to selectively bind and activate ER thereby modulating ER -mediated signal transduction DPN exerts its biological activity primarily through selective agonistic activation of ER DPN demonstrates high binding affinity with an EC50 value of approximately 0 85 nM Based on these pharmacological properties DPN holds research potential in neuroprotection and the study of neuropathological conditions
Catalog No. 50-000-89286 Supplier Apexbio Technology LLC Supplier No. B67635
DPN (1428-67-7) is a non-steroidal agonist targeting estrogen receptor (ER ) It is designed to selectively bind and activate ER thereby modulating ER -mediated signal transduction DPN exerts its biological activity primarily through selective agonistic activation of ER DPN demonstrates high binding affinity with an EC50 value of approximately 0 85 nM Based on these pharmacological properties DPN holds research potential in neuroprotection and the study of neuropathological conditions