Learn More
Medchemexpress LLC DPN (Diarylpropionitrile) | 1428-67-7 | MFCD01695412 | 239.27 g/mol | C15H13NO2 | 50MG
SDP
Catalog No. 5000238002
Encompass_Preferred
DPN (Diarylpropionitrile) is a non-steroidal, selective estrogen receptor β (ERβ) ligand used in research to probe ERβ signaling and neuroprotective mechanisms. It is commonly used in in vitro and in vivo pharmacology studies investigating estrogen receptor function and neurobiology.
Catalog No. 50-002-38002 Supplier Medchemexpress LLC Supplier No. HY1245250MG
DPN (Diarylpropionitrile) is a non-steroidal, selective estrogen receptor β (ERβ) ligand used in research to probe ERβ signaling and neuroprotective mechanisms. It is commonly used in in vitro and in vivo pharmacology studies investigating estrogen receptor function and neurobiology.