Learn More
Apexbio Technology LLC Dpn I(Synonyms: DpnI, DpnI Restriction Enzyme, Dam methylation-sensitive restriction enzyme DpnI), 5000U.
SDP
Catalog No. 5000061253
Encompass_Preferred
Dpn I is a restriction endonuclease sensitive to Dam methylation specifically recognizing and cleaving the adenine N6-methylated sequence GA/TC within DNA Cleavage is dependent on adenine methylation status mammalian CpG methylation overlapping Dpn I sites blocks cleavage and methylation by Dcm is not recognized by this enzyme It is routinely used for the rapid digestion of plasmid DNA PCR fragments and genomic DNA and finds broad utility in genetic typing molecular cloning Southern blotting SNP genotyping and restriction fragment length polymorphism (RFLP) analyses
Catalog No. 50-000-61253 Supplier Apexbio Technology LLC Supplier No. K11045000
Dpn I is a restriction endonuclease sensitive to Dam methylation specifically recognizing and cleaving the adenine N6-methylated sequence GA/TC within DNA Cleavage is dependent on adenine methylation status mammalian CpG methylation overlapping Dpn I sites blocks cleavage and methylation by Dcm is not recognized by this enzyme It is routinely used for the rapid digestion of plasmid DNA PCR fragments and genomic DNA and finds broad utility in genetic typing molecular cloning Southern blotting SNP genotyping and restriction fragment length polymorphism (RFLP) analyses