Learn More
Agilent Technologies DPN I RESTRICTION ENZYME
Catalog No. 50125830
Encompass
Dpn I Restriction Enzyme, is negative for nonspecific nucleases. It is used when cloning into multiple cloning site of pBluescript II phagemid vector. It includes an optimal (10X) buffer and/or a 10X concentrate of Universal buffer.
Catalog No. 50-125-830 Supplier Agilent Technologies Supplier No. 500402
Dpn I Restriction Enzyme, is negative for nonspecific nucleases. It is used when cloning into multiple cloning site of pBluescript II phagemid vector. It includes an optimal (10X) buffer and/or a 10X concentrate of Universal buffer.