Learn More
New England Biolabs, Inc. DpnI – 5000 units
SDP
Catalog No. 50812090
Encompass
A restriction endonuclease that recognizes the sequence GA_^TC.
  • DpnI cleaves only when its recognition site is methylated
  • Cleavage of mammalian genomic DNA is blocked by overlapping CpG methylation
  • Methylation-sensitive restriction enzyme
  • Time-Saver qualified for digestion in 5-15 minutes
  • 100% activity in rCutSmart Buffer (over 210 enzymes are available in the same buffer)
Catalog No. 50-812-090 Supplier New England Biolabs, Inc. Supplier No. R0176L
A restriction endonuclease that recognizes the sequence GA_^TC.
  • DpnI cleaves only when its recognition site is methylated
  • Cleavage of mammalian genomic DNA is blocked by overlapping CpG methylation
  • Methylation-sensitive restriction enzyme
  • Time-Saver qualified for digestion in 5-15 minutes
  • 100% activity in rCutSmart Buffer (over 210 enzymes are available in the same buffer)