Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPNI-caged-GABA, Tocris Bioscience™
SDP

Catalog No. 299110
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-911-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 29-911-0 Supplier R&D Systems Supplier No. 2991/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nitroindoline-caged GABA

Nitroindoline-caged GABA (Cat.No. 0344) with similar photochemical properties to MNI-glutamate (Cat. No. 1490); for example, the same quantum yield (0.085). Highly water soluble; exhibits fast photorelease that is efficient at near-UV and 405 nm wavelengths.

Chemical Identifiers

CAS 927866-58-8
Molecular Formula C15H23N3O12P2
Molecular Weight (g/mol) 499.306
InChI Key QLCHBTSDKGMLRN-UHFFFAOYSA-N
Synonym dpni-caged-gaba, 1-4-aminobutanoyl-4-1,3-bis dihydroxyphosphoryloxy propan-2-yloxy-7-nitroindoline, 4-amino-1-2,3-dihydro-7-nitro-4-2-phosphonooxy-1-phosphonooxy methyl ethoxy-1h-indol-1-yl-1-butanone
PubChem CID 15953814
IUPAC Name [2-[[1-(4-aminobutanoyl)-7-nitro-2,3-dihydroindol-4-yl]oxy]-3-phosphonooxypropyl] dihydrogen phosphate
SMILES C1CN(C2=C(C=CC(=C21)OC(COP(=O)(O)O)COP(=O)(O)O)[N+](=O)[O-])C(=O)CCCN

Specifications

Quantity 10 mg
Formula Weight 499.3
Percent Purity >98%
Chemical Name or Material DPNI-caged-GABA
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.