Learn More
New England Biolabs, Inc. DpnII – 1000 units
SDP
Catalog No. 50812216
Encompass
A restriction endonuclease that recognizes the sequence ^GATC_.
Catalog No. 50-812-216 Supplier New England Biolabs, Inc. Supplier No. R0543S
A restriction endonuclease that recognizes the sequence ^GATC_.