Learn More
New England Biolabs, Inc. DpnII – 5000 units
SDP
Catalog No. 50812214
Encompass
A restriction endonuclease that recognizes the sequence ^GATC_.
Catalog No. 50-812-214 Supplier New England Biolabs, Inc. Supplier No. R0543L
A restriction endonuclease that recognizes the sequence ^GATC_.