Learn More
New England Biolabs, Inc. DpnII – 1000 units
SDP
Catalog No. 50812217
Encompass
A restriction endonuclease that recognizes the sequence ^GATC_.
Catalog No. 50-812-217 Supplier New England Biolabs, Inc. Supplier No. R0543T
A restriction endonuclease that recognizes the sequence ^GATC_.