Learn More
New England Biolabs, Inc. DpnII – 5000 units
SDP
Catalog No. 50812215
Encompass
A restriction endonuclease that recognizes the sequence ^GATC_.
Catalog No. 50-812-215 Supplier New England Biolabs, Inc. Supplier No. R0543M
A restriction endonuclease that recognizes the sequence ^GATC_.