Learn More
Cayman Chemical DPP-9 Inhibitor 42, Purity: ≥95%, 50 mg
SDP
Catalog No. 502942774
Encompass_Preferred
A DPP-9 inhibitor (IC50 = 0.0034 µM); selective for DDP-9 over DPP-8, DPP-4, DPP-2, FAP, and prolyl endopeptidase (IC50s = 0.6, >10, >5, 10, and >10 µM, respectively); induces LDH release from THP-1 and HL-60 leukemia cells at 10 µM
Catalog No. 50-294-2774 Supplier Cayman Chemical Supplier No. 3988150 MG
A DPP-9 inhibitor (IC50 = 0.0034 µM); selective for DDP-9 over DPP-8, DPP-4, DPP-2, FAP, and prolyl endopeptidase (IC50s = 0.6, >10, >5, 10, and >10 µM, respectively); induces LDH release from THP-1 and HL-60 leukemia cells at 10 µM