Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chembio Diagnostic Systems, Inc. DPP HIV-Syphilis Control Pack
GSA_VASDP

Catalog No. 23999114
Change view
Click to view available options
Quantity:
1 Set of Controls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-999-114 1 Set of Controls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-999-114 Supplier Chembio Diagnostic Systems, Inc. Supplier No. 6095550
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reactive/Nonreactive Controls are quality control reagents for use with DPP HIV-Syphilis.

  • Chembio's DPP HIV-Syphilis Reactive/Nonreactive Controls are quality control reagents for use with DPP HIV-Syphilis.
  • Each DPP HIV-Syphilis Rapid Test Control Pack contains a Product Insert and three 3 Vials
  • One HIV-1 / Syphilis Reactive Control
  • One HIV-2 Reactive Control
  • One Non-reactive Control

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance Moderately Complex
Content And Storage 2°C to 8°C
Control Sets One HIV-1 / Syphilis Reactive Control, One HIV-2 Reactive Control, One Non-reactive Control
CE Marker Yes
Clia Complexity Moderate
Detectable Analytes HIV, T. pallidum
Sample Type Fingerstick whole blood, venous whole blood, or plasma
Type Assay Controls
For Use With (Application) Assay Controls
Material Polypropylene Vials
Packaging Type 1 kit (contains 3 vials)
Quantity 1 Set of Controls
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.