Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chembio Diagnostic Systems, Inc.™ DPP HIV-Syphilis PROMO
SDP

Catalog No. 23999120
Change view
Click to view available options
Includes:
20 Kits and 1 Reader
35 Kits and 2 Readers
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
23-999-120 35 Kits and 2 Readers
23-999-119 20 Kits and 1 Reader
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-999-120 Supplier Chembio Diagnostic Systems, Inc.™ Supplier No. 10001022
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The first and only dual rapid HIV-Syphilis test in the US, representing a significant step forward in rapid testing for sexually transmitted diseases.

  • Test utilizes patented Dual Path Platform (DPP) technology, providing results in 15 minutes
  • Easy to use and can be stored at room temperature
  • Single-use rapid, qualitative, multiplex, immunoassay for the detection of antibodies to HIV-1/2, and/or Treponema pallidum bacteria (the causative agent of syphilis)
  • Patented DPP technology allows for higher sensitivity: >99% for HIV and >94% for T. pallidum
  • Only requires 10 μL of either fingerstick whole blood, venous whole blood or plasma
  • Provides results in 15 minutes and is read with the DPP Micro Reader

Specifications

Content And Storage 2°C to 25°C
CE Marker Yes
Clia Complexity Waived
Detectable Analytes HIV, T. pallidum
Sample Type Fingerstick Whole Blood, Venous Whole Blood, Plasma
Type Rapid Test
For Use With (Application) DPP Rapid Test
Includes 35 Kits and 2 Readers
Packaging Type Kit
Quantity 20 Tests/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.