Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chembio Diagnostic Systems, Inc. DPP HIV-Syphilis
GSA_VASDP

Catalog No. 23999113
Change view
Click to view available options
Quantity:
20 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-999-113 20 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-999-113 Supplier Chembio Diagnostic Systems, Inc. Supplier No. 6595020
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The first and only dual rapid HIV-Syphilis test in the US and represents a significant step forward in rapid testing for sexually transmitted diseases.

  • This test utilizes our patented Dual Path Platform (DPP) technology, provides results in 15 minutes, is easy to use and can be stored at room temperature.
  • This test is a single-use rapid, qualitative, multiplex, immunoassay for the detection of antibodies to HIV-1/2, and/or Treponema pallidum bacteria (the causative agent of syphilis) in fingerstick whole blood, venous whole blood or plasma specimens.
  • Patented DPP technology allows for higher sensitivity: >99% for HIV and >94% for T. pallidum
  • Only requires 10 μL of either fingerstick whole blood, venous whole blood or plasma
  • Provides results in 15 minutes and is read with the DPP Micro Reader
  • Combined reimbursement: CPT Codes 86703, 86780

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance CLIA Waived
Content And Storage 2°C to 25°C
Control Sets N/A - test device kit
CE Marker Yes
Clia Complexity Waived
Detectable Analytes HIV, T. pallidum
Sample Type Fingerstick whole blood, venous whole blood, or plasma
For Use With (Application) Chembio DPP Rapid Test
Packaging Type Kit
Quantity 20 Tests/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.