Learn More
Medchemexpress LLC HY-108319 50mg Medchemexpress, DPP-IV-IN-2 CAS:136259-18-2 Purity:>98%
SDP
Catalog No. 502080738
Encompass_Preferred
Medchemexpress, HY-108319 50mg DPP-IV-IN-2 CAS:136259-18-2 DPP-IV-IN-2 is an inhibitor of both dipeptidyl peptidase IV (DPIV) and DP8/9 with IC50s of 0.1 and 0.95 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0738 Supplier Medchemexpress LLC Supplier No. HY10831950MG
Medchemexpress, HY-108319 50mg DPP-IV-IN-2 CAS:136259-18-2 DPP-IV-IN-2 is an inhibitor of both dipeptidyl peptidase IV (DPIV) and DP8/9 with IC50s of 0.1 and 0.95 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.