Learn More
MilliporeSigma DPP-IV INHIBITOR 1ML
Catalog No. NC9010908
Encompass
Inhibitor, DPP-IV; 1mL; Reagent inhibits degradation by the DPP IV enzyme; Storage: 4 deg.C and -20 deg.C for long term storage (> 2 weeks)
Catalog No. NC9010908 Supplier MilliporeSigma Supplier No. DPP4
Inhibitor, DPP-IV; 1mL; Reagent inhibits degradation by the DPP IV enzyme; Storage: 4 deg.C and -20 deg.C for long term storage (> 2 weeks)