Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chembio DPP™ Zika IgM Assay System
GSA_VASDP

Catalog No. 23999106
Change view
Click to view available options
Quantity:
20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-999-106 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-999-106 Supplier Chembio Supplier No. 6595550
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

20 Individually Pouched DPP™ Zika IgM Test Devices, 20 Disposable 10μL Microsafe™ Tubes, 20 Sample Vials, 20 Transfer Pipets (100μL), 1 DPP IgM Sample Buffer, Blue Cap, 1 DPP IgM Running Buffer, Yellow Cap, 1 Product Insert, 1 Quick Reference Guide, Fact sheet for patients, Fact sheet for Health care workers

The 15-minute rapid test for detecting Zika virus infection

For the detection of Zika virus IgM antibodies in fingerstick whole blood, EDTA venous whole blood, EDTA plasma and serum.

  • Fast: Provides results in as little as 15 minutes
  • Flexible: Detects Zika virus IgM antibodies from 8 days up to 12 weeks
  • Easy to Use: Provides objective results using simple, handheld digital reader (DPP™ Micro Reader)
  • Received FDA Emergency Use Authorization (EUA)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Spécifications

Content And Storage 2°C to 8°C
Detectable Analytes Zika Virus IgM
Type Rapid Test
For Use With (Application) Detection of Zika virus infection in Fingerstick Whole Blood, Venous Whole blood, Serum, Plasma
Includes 20 Individually Pouched DPP™ Zika IgM Test Devices, 20 Disposable 10μL Microsafe™ Tubes, 20 Sample Vials, 20 Transfer Pipets (100μL), 1 DPP IgM Sample Buffer, Blue Cap, 1 DPP IgM Running Buffer, Yellow Cap, 1 Product Insert, 1 Quick Reference Guide, Fact sheet for patients, Fact sheet for Health care workers
Quantity 20
Test Time 15 min.

  1. The test has not been FDA cleared or approved
  2. The test has been authorized by FDA under an EUA for use by authorized laboratories
  3. The text has been authorized only for the diagnosis of Zika virus infection and not for any other viruses or pathogens
  4. The test is only authorized for the duration of the declaration that circumstances exist justifying the authorization of the emergency use of in vitro diagnostic tests for detection of the Zika virus and/or diagnosis of Zika virus infection under section 564(b)(1) of the Act, 21 U.S.C. § 360bbb-3(b)(1), unless the authorization is terminated or revoked sooner.

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.