Learn More
Aviva Systems Biology Corp DPP4 Antibody - C-terminal region (ARP63319_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63319_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DPP4. It was validated on Western Blot. . Alternate names: ADABP, ADCP2, CD26, DPPIV, TP103ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-938