Learn More
Aviva Systems Biology Corp DPP6 Antibody - middle region (ARP44867_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44867_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DPP6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DPP6. Alternate names: DPPX, MGC46605, VF2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58921