Learn More
Enzo Life Sciences DPP8 (human), (recombinant) (GST-tag) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE5270010
Encompass_Preferred
Alternative name: Dipeptidyl peptidase 8. Purity: ≥70% (SDS-PAGE). Formulation: Liquid. In 40mM TRIS-HCl, pH 8.0, containing 110mM sodium chloride, 2.2mM potassium chloride, and 20% glycerol. Source: Produced in insect cells. Active recombinant human DPP8 is fused at the N-terminus to a GST-tag. Produced in a baculovirus expression system. Long Term Storage: -80°C.
Catalog No. 50-201-2068