Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DPP8 protein, human (sf9) | >90.0% | 10UG
SDP

Catalog No. 5000176782
Encompass_Preferred

Recombinant full-length human DPP8 protein expressed in Sf9 insect cells and supplied tag-free. The protein exhibits dipeptidyl peptidase activity, cleaving N-terminal dipeptides from substrates with proline or alanine at position 2, and participates in suppression of NLRP1 and CARD8-mediated inflammasome activation.

  • Full-length human sequence (M1-I898).
  • Expressed in Sf9 insect cells.
  • Tag-free preparation.
  • Purity >90% by reducing SDS-PAGE.
  • Molecular weight ~103.4 kDa.
  • Supplied in 50 mM HEPES, 200 mM NaCl, 20% glycerol, 1 mM DTT, pH 7.5.
  • Store at -80°C; stable at -20°C for 3 months after opening; avoid repeated freeze-thaw.
  • Shipped on dry ice.

Catalog No. 50-001-76782 Supplier Medchemexpress LLC Supplier No. HYP70187310UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant full-length human DPP8 protein expressed in Sf9 insect cells and supplied tag-free. The protein exhibits dipeptidyl peptidase activity, cleaving N-terminal dipeptides from substrates with proline or alanine at position 2, and participates in suppression of NLRP1 and CARD8-mediated inflammasome activation.

  • Full-length human sequence (M1-I898).
  • Expressed in Sf9 insect cells.
  • Tag-free preparation.
  • Purity >90% by reducing SDS-PAGE.
  • Molecular weight ~103.4 kDa.
  • Supplied in 50 mM HEPES, 200 mM NaCl, 20% glycerol, 1 mM DTT, pH 7.5.
  • Store at -80°C; stable at -20°C for 3 months after opening; avoid repeated freeze-thaw.
  • Shipped on dry ice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.