Learn More
Medchemexpress LLC DPP8 protein, human (sf9, His) | 97.0% | 100UG
SDP
Catalog No. 5000176659
Encompass_Preferred
Recombinant human DPP8 protein expressed in Sf9 insect cells with an N-terminal 8xHis tag. Supplied as a filtered, buffered solution suitable for research use and shipped on dry ice.
Catalog No. 50-001-76659 Supplier Medchemexpress LLC Supplier No. HYP701874100UG
Recombinant human DPP8 protein expressed in Sf9 insect cells with an N-terminal 8xHis tag. Supplied as a filtered, buffered solution suitable for research use and shipped on dry ice.