Learn More
Medchemexpress LLC DPP8 protein, human (Sf9, His) | 97.0% | 10UG
SDP
Catalog No. 5000175734
Encompass_Preferred
Recombinant human DPP8 protein produced in Sf9 insect cells with an N-terminal 8xHis tag. Supplied as a concentrated solution suitable for enzymatic assays, structural studies, and antibody validation; accession Q6V1X1 denotes the canonical human sequence.
  • Recombinant human DPP8 expressed in Sf9 insect cells.
  • N-terminal 8xHis tag for purification and detection.
  • High purity (~97% by reducing SDS-PAGE).
  • Approximate molecular weight ~100 kDa.
  • Available in small research sizes (10 μg, 50 μg, 100 μg).
Catalog No. 50-001-75734 Supplier Medchemexpress LLC Supplier No. HYP70187410UG
Recombinant human DPP8 protein produced in Sf9 insect cells with an N-terminal 8xHis tag. Supplied as a concentrated solution suitable for enzymatic assays, structural studies, and antibody validation; accession Q6V1X1 denotes the canonical human sequence.
  • Recombinant human DPP8 expressed in Sf9 insect cells.
  • N-terminal 8xHis tag for purification and detection.
  • High purity (~97% by reducing SDS-PAGE).
  • Approximate molecular weight ~100 kDa.
  • Available in small research sizes (10 μg, 50 μg, 100 μg).