Learn More
Enzo Life Sciences Phosphatidic acid, dipalmitoyl (100mg). CAS: 169051-60-9
SDP
Supplier:  Enzo Life Sciences BMLLP1030100
Encompass_Preferred
A mitogenic lipid phosphate produced via the action of phospholipase D. Mediates agonist-dependent Raf-1 translocation and activation of the MAP kinase cascade. Alternative name: 1,2-Dipalmitoyl-sn-glycero-3-phosphate . sodium salt, DPPA. Purity: ≥98% (TLC). Solubility: Fine suspension in ethanol (5mg/ml) with 15 minutes of sonication at room temperature. Long Term Storage: -20°C.
Catalog No. 50-201-1155