Learn More
Cayman Chemical (D)-PPA 1 (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502911378
Encompass_Preferred
An inhibitor of the PD-1-PD-L1 protein-protein interaction; binds to the extracellular domain of PD-L1 (Kd = 0.51 µM for the recombinant human protein); decreases tumor volume and improves survival in a CT26 murine colorectal cancer model at 2 mg/kg
Catalog No. 50-291-1378 Supplier Cayman Chemical Supplier No. 3564710 MG
An inhibitor of the PD-1-PD-L1 protein-protein interaction; binds to the extracellular domain of PD-L1 (Kd = 0.51 µM for the recombinant human protein); decreases tumor volume and improves survival in a CT26 murine colorectal cancer model at 2 mg/kg