Learn More
Echelon Biosciences Research Labs DPPA 250 mg
SDP
Supplier:  Echelon Biosciences Research Labs L4116250MG
Encompass
1,2-Dipalmitoyl-sn-glycero-3-phosphate, sodium salt (DPPA) is a form of phosphatidic acid (PA), a major component of cell membranes with important biophysical, metabolic, and signaling roles. PA has been implicated in many aspects of animal cell biochemistry and physiology, including cell proliferation and differentiation, cell transformation, tumor progression and survival signaling. This item is non returnable
Catalog No. 50-720-512