Learn More
Aviva Systems Biology Corp DPPA2 Antibody - N-terminal region (ARP52662_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52662_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DPPA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DPPA2. Alternate names: PESCRG1, CT100, ECAT15-2ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-60798