Learn More
Aviva Systems Biology Corp DPPA4 Antibody - N-terminal region (ARP57139_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57139_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DPPA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DPPA4. Alternate names: 2410091M23Rik, FLJ10713ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53963