Learn More
Apexbio Technology LLC 1 2-Dipalmitoyl-sn-glycero-3-PC 100mg
SDP
Catalog No. 503107431
Encompass_Preferred
1 2-Dipalmitoyl-sn-glycero-3-phosphorylcholine (DPPC) is a zwitterionic glycerophospholipid commonly used to form lipid monolayers bilayers and liposomes for various applications DPPC-liposomes effectively serve as delivery vehicles to induce immune responses against GSL antigens in mice
Catalog No. 50-310-7431 Supplier Apexbio Technology LLC Supplier No. C4002250
1 2-Dipalmitoyl-sn-glycero-3-phosphorylcholine (DPPC) is a zwitterionic glycerophospholipid commonly used to form lipid monolayers bilayers and liposomes for various applications DPPC-liposomes effectively serve as delivery vehicles to induce immune responses against GSL antigens in mice