Learn More
Kyfora Bio DPPC 500 MG
Catalog No. 502559775
Encompass_Preferred
DPPC (1,2-Dipalmitoyl-sn-glycerol-3-Phosphocholine), a variant of phosphatidylcholine, is an amphipathic phospholipid used for gene and drug delivery formulations. DPPC has been shown to have excellent biodegradability and drug encapsulation capacity, and when combined with other lipids has high cellular uptake efficiency, making it ideal for use in liposomal formulations. DPPC is also an integral element of pulmonary surfactants and is often used when studying bilayers found in the human body.Key application(s): LNP, drug delivery
Catalog No. 50-255-9775 Supplier Kyfora Bio Supplier No. 26449500
DPPC (1,2-Dipalmitoyl-sn-glycerol-3-Phosphocholine), a variant of phosphatidylcholine, is an amphipathic phospholipid used for gene and drug delivery formulations. DPPC has been shown to have excellent biodegradability and drug encapsulation capacity, and when combined with other lipids has high cellular uptake efficiency, making it ideal for use in liposomal formulations. DPPC is also an integral element of pulmonary surfactants and is often used when studying bilayers found in the human body.Key application(s): LNP, drug delivery