Learn More
Medchemexpress LLC DPPC-d62 | 25582-63-2 | 99.0% | 796.42 | 50 MG
SDP
Catalog No. 5000345345
Encompass_Preferred
DPPC-d62 is deuterium labeled DPPC. DPPC is a zwitterionic phosphoglyceride that can be used for the preparation of liposomal monolayers. DPPC-liposome can effectively serve as a delivery vehicle for inducing immune responses against GSL antigen in mice.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-45345 Supplier Medchemexpress LLC Supplier No. HY109506S1MG
DPPC-d62 is deuterium labeled DPPC. DPPC is a zwitterionic phosphoglyceride that can be used for the preparation of liposomal monolayers. DPPC-liposome can effectively serve as a delivery vehicle for inducing immune responses against GSL antigen in mice.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.