Learn More
Medchemexpress LLC DPPC-d9 (Synonyms: 129Y83-d9) | 63-89-8 | 95.0% | 743.09 | 100 MG
SDP
Catalog No. 5000345933
Encompass_Preferred
DPPC-d9 is a deuterium-labeled DPPC. DPPC is a zwitterionic phosphoglyceride used for preparing liposomal monolayers. DPPC-liposome effectively serves as a delivery vehicle for inducing immune responses against GSL antigen in mice.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-45933 Supplier Medchemexpress LLC Supplier No. HY109506S11MG
DPPC-d9 is a deuterium-labeled DPPC. DPPC is a zwitterionic phosphoglyceride used for preparing liposomal monolayers. DPPC-liposome effectively serves as a delivery vehicle for inducing immune responses against GSL antigen in mice.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, largely as tracers for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.