Learn More
Echelon Biosciences Research Labs DPPE 250 mg
SDP
Catalog No. 50720503
Encompass
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine (DPPE) is a water soluble derivative of phosphatidylethanolamine with (16:0) palmitoyl acyl chains. Phosphatidylethanolamine (PE) is formed primarily in the reaction of CDP-ethanolamine and diacylglycerol. PE is one of the major components of cell membranes in bacteria and in nervous system. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-503 Supplier Echelon Biosciences Research Labs Supplier No. L2116250MG
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine (DPPE) is a water soluble derivative of phosphatidylethanolamine with (16:0) palmitoyl acyl chains. Phosphatidylethanolamine (PE) is formed primarily in the reaction of CDP-ethanolamine and diacylglycerol. PE is one of the major components of cell membranes in bacteria and in nervous system. This item is non returnable