Learn More
Selleck Chemical LLC DPPE fumarate-E0376-5MG
SDP
Catalog No. 503136664
Encompass_Preferred
DPPE fumarate (Tesmilifene fumarate) is a novel potentiator of chemotherapy which is found to potentiate a wide range of cytotoxics and to protect normal cells
Catalog No. 50-313-6664 Supplier Selleck Chemical LLC Supplier No. E03765MG
DPPE fumarate (Tesmilifene fumarate) is a novel potentiator of chemotherapy which is found to potentiate a wide range of cytotoxics and to protect normal cells