Learn More
Cayman Chemical DPPE-MPEG(2000) (ammonium salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502949940
Encompass_Preferred
A PEGylated form of DPPE; has been used in the synthesis of anionic liposomes for drug redistribution and toxicity prevention
Catalog No. 50-294-9940 Supplier Cayman Chemical Supplier No. 3465010 MG
A PEGylated form of DPPE; has been used in the synthesis of anionic liposomes for drug redistribution and toxicity prevention