Learn More
Cayman Chemical DPPE-MPEG 2000 25mg
SDP
Catalog No. 502946665
Encompass_Preferred
A PEGylated form of DPPE; has been used in the synthesis of anionic liposomes for drug redistribution and toxicity prevention
Catalog No. 50-294-6665 Supplier Cayman Chemical Supplier No. 3465025 MG
A PEGylated form of DPPE; has been used in the synthesis of anionic liposomes for drug redistribution and toxicity prevention