Learn More
Cayman Chemical DPPE-MPEG 2000 5mg
SDP
Catalog No. 502949852
Encompass_Preferred
A PEGylated form of DPPE; has been used in the synthesis of anionic liposomes for drug redistribution and toxicity prevention
Catalog No. 50-294-9852 Supplier Cayman Chemical Supplier No. 346505 MG
A PEGylated form of DPPE; has been used in the synthesis of anionic liposomes for drug redistribution and toxicity prevention