Learn More
Medchemexpress LLC DPPF (1,1'-Ferrocenediyl-bis(diphenylphosphine)) | 12150-46-8 | 554.38 | 25 G
SDP
Catalog No. 5000450035
Encompass_Preferred
DPPF (1,1'-Ferrocenediyl-bis(diphenylphosphine)) is a metal ligand that enhances the stability of metal complexes. It is suitable for use as a laboratory chemical in the manufacture of substances and for research in ovarian cancer.
  • Enhances the stability of metal complexes
  • Functions as a metal ligand
  • Applicable in ovarian cancer research
  • Used as a biochemical assay reagent
Catalog No. 50-004-50035 Supplier Medchemexpress LLC Supplier No. 25GHYW001950
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
DPPF (1,1'-Ferrocenediyl-bis(diphenylphosphine)) is a metal ligand that enhances the stability of metal complexes. It is suitable for use as a laboratory chemical in the manufacture of substances and for research in ovarian cancer.
  • Enhances the stability of metal complexes
  • Functions as a metal ligand
  • Applicable in ovarian cancer research
  • Used as a biochemical assay reagent