Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dppf Pd G3

Catalog No. 8049831G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-498-31G 250 mg
80-498-3250MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 80-498-31G Supplier Sigma Aldrich Supplier No. 8049831G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dppf Pd G3

Specifications

CAS 2903-48-2
Melting Point 134°C to 208°C (MP data is all decomp.)
Molecular Formula C47H41FeNO3P2PdS
Linear Formula C47H41FeNO3P2PdS
Quantity 250 mg
MDL Number MFCD28009174
Synonym Methanesulfonato 1,1-ferrocenediyl-bis(diphenylphosphino) (2'-amino-1,1'-biphenyl-2-yl) palladium(II)
Molecular Weight (g/mol) 924.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.