Learn More
Echelon Biosciences Research Labs DPPG 1 g
SDP
Catalog No. 50720517
Encompass
1,2-Dipalmitoyl-sn-glycero-3-phosphoglycerol, sodium salt (DPPG) is a form of phosphatidylglycerol (PG), a minor component (1-2%) of most of cell membranes. It can be found at higher concentrations (up to 11% of the total lipid content) in lung surfactant. Its presence in amniotic fluid indicates fetal lung maturity and is the basis of clinical testing for fetal fitness. PG is also used by cells as an intermediate in cardiolipin synthesis. Bacterial membranes contain up to 20% of PG. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-517 Supplier Echelon Biosciences Research Labs Supplier No. L51161G
1,2-Dipalmitoyl-sn-glycero-3-phosphoglycerol, sodium salt (DPPG) is a form of phosphatidylglycerol (PG), a minor component (1-2%) of most of cell membranes. It can be found at higher concentrations (up to 11% of the total lipid content) in lung surfactant. Its presence in amniotic fluid indicates fetal lung maturity and is the basis of clinical testing for fetal fitness. PG is also used by cells as an intermediate in cardiolipin synthesis. Bacterial membranes contain up to 20% of PG. This item is non returnable