Learn More
Dojindo Molecular Technologies Inc DPPH Antioxidant Assay Kit enables quick and easy measurements of the antioxidant capacity of a sample and a reproducible detection of antioxidant capacity assay 100 tests
SDP
Catalog No. NC1933295
Encompass_Preferred
Recent findings suggest that a decline in internal antioxidant capacity causes the onset of various diseases and health impairment. Consequently, interest in antioxidant-rich foods has been recently increasing. Shimamura et al. improved the 2,2-diphenyl-1-picrylhydrazyl (DPPH) assay, a procedure for evaluating antioxidant capacities, and reported it may be applied as a standard method to evaluate the antioxidant capacity of antioxidants. The DPPH Antioxidant Assay Kit is based on the DPPH assay improved by Shimamura and enables quick and easy measurements of the antioxidant capacity of a sample. Using this kit, the antioxidant capacity is expressed as the Trolox equivalent antioxidant capacity (TEAC), a value calculated from the IC50 of the antioxidant and the IC50 of Trolox. The DPPH Reagent and Trolox Standard included in this kit only need to be dissolved prior to use. The protocol for this kit supports a microplate assay format for the simultaneous analysis of multiple samples.
Catalog No. NC1933295 Supplier Dojindo Molecular Technologies Inc Supplier No. D67810
Recent findings suggest that a decline in internal antioxidant capacity causes the onset of various diseases and health impairment. Consequently, interest in antioxidant-rich foods has been recently increasing. Shimamura et al. improved the 2,2-diphenyl-1-picrylhydrazyl (DPPH) assay, a procedure for evaluating antioxidant capacities, and reported it may be applied as a standard method to evaluate the antioxidant capacity of antioxidants. The DPPH Antioxidant Assay Kit is based on the DPPH assay improved by Shimamura and enables quick and easy measurements of the antioxidant capacity of a sample. Using this kit, the antioxidant capacity is expressed as the Trolox equivalent antioxidant capacity (TEAC), a value calculated from the IC50 of the antioxidant and the IC50 of Trolox. The DPPH Reagent and Trolox Standard included in this kit only need to be dissolved prior to use. The protocol for this kit supports a microplate assay format for the simultaneous analysis of multiple samples.