Learn More
Enzo Life Sciences DPPIV/CD26 Assay kit for biological samples (1x96wells)
SDP
Catalog No. NC9058013
Encompass_Preferred
Because DPPIV/CD26 protein levels may not accurately reflect the levels of active DPPIV enzyme, it is useful to measure DPPIV activity rather than performing DPPIV immunoassay. The DPPIV/CD26 Assay kit for biological samples is a complete assay system designed to measure DPPIV activity in biological fluids such as plasma, serum, urine, and saliva. Uses for the kit include correlation of DPPIV activity with disease states or determination of the efficacy of DPPIV inhibitors administered .½-volume NBS white microplate (Prod. No. BML-KI571)
Catalog No. NC9058013 Supplier Enzo Life Sciences Supplier No. BMLAK4980001
Because DPPIV/CD26 protein levels may not accurately reflect the levels of active DPPIV enzyme, it is useful to measure DPPIV activity rather than performing DPPIV immunoassay. The DPPIV/CD26 Assay kit for biological samples is a complete assay system designed to measure DPPIV activity in biological fluids such as plasma, serum, urine, and saliva. Uses for the kit include correlation of DPPIV activity with disease states or determination of the efficacy of DPPIV inhibitors administered .½-volume NBS white microplate (Prod. No. BML-KI571)