Learn More
Genscript Corporation DPPIV/CD26 hFc Chimera Human
Catalog No. 5000058603
Encompass
CD26/dipeptidyl peptidase (DPP)IV is a membrane-bound protein found in many cell types of the body and a soluble form is present in body fluids There is longstanding evidence that various primary tumors and also metastases express DPPIV/CD26 to a variable extent
Catalog No. 50-000-58603 Supplier Genscript Corporation Supplier No. Z04249500
CD26/dipeptidyl peptidase (DPP)IV is a membrane-bound protein found in many cell types of the body and a soluble form is present in body fluids There is longstanding evidence that various primary tumors and also metastases express DPPIV/CD26 to a variable extent